Learn More
HELIX CHROMATOGRAPHY Coresep SB 4.6 x 150 mm, 2.7 um
Catalog No. NC2233385
Encompass
Core-shell mixed-mode stationary phase combining reversed-phase with strong anion-exchange and cation-exclusion mechanisms. Provides retention for hydrophobic drugs, acidic counter-ions, weak acids, and basic polymers. Particularly effective for separations requiring strong anion-exchange interactions and high-efficiency performance.Single-ligand design with a surface-proximal proprietary basic group that shields residual silanols and reduces secondary interactions.Incorporates surface-bound ion-pairing functionality, enabling full MS compatibility without the need for mobile phase ion-pairing reagents.
This item is not returnable. View return policy
Catalog No. NC2233385 Supplier HELIX CHROMATOGRAPHY Supplier No. CSB461529
Core-shell mixed-mode stationary phase combining reversed-phase with strong anion-exchange and cation-exclusion mechanisms. Provides retention for hydrophobic drugs, acidic counter-ions, weak acids, and basic polymers. Particularly effective for separations requiring strong anion-exchange interactions and high-efficiency performance.Single-ligand design with a surface-proximal proprietary basic group that shields residual silanols and reduces secondary interactions.Incorporates surface-bound ion-pairing functionality, enabling full MS compatibility without the need for mobile phase ion-pairing reagents.