Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Corey lactone benzoate, Thermo Scientific™

Catalog No. AC233520010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC233520010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC233520010 Supplier Thermo Scientific Chemicals Supplier No. 233520010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39746-00-4
Infrared Spectrum Authentic
Molecular Formula C15H16O5
Quantity 1g
MDL Number MFCD00674102
Beilstein 18, V,2, 499
Specific Rotation Condition −80° (25°C c=0.8,CHCl3)
Specific Rotation -76° to -84° (25°C, 589nm) (c=0.8, CHCl3)
Synonym --corey lactone benzoate, corey lactone benzoate, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxohexahydro-2h-cyclopenta b furan-5-yl benzoate, 3ar,4s,5r,6as-5-benzoyloxy hexahydro-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 1s,5r,6s,7r-7-benzoyloxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as-5-benzoyloxy hexahydro-4-hydroxymethyl-2h-cyclopentafuran-2-one, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-5-yl benzoate, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxo-hexahydrocyclopenta b furan-5-yl benzoate, benzoyl-corey lactone, pubchem19127
InChI Key OBRRYUZUDKVCOO-FVCCEPFGSA-N
SMILES C1C2C(CC(=O)O2)C(C1OC(=O)C3=CC=CC=C3)CO
IUPAC Name [(3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] benzoate
Molecular Weight (g/mol) 276.288
PubChem CID 2724454
Formula Weight 276.29
Percent Purity ≥97.5%
Chemical Name or Material (-)-Corey lactone benzoate
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.