Learn More
Medchemexpress LLC HY-W008393 100mg Medchemexpress, (-)-Corey lactone diol CAS:32233-40-2 Purity:>98%
SDP
Catalog No. 501934514
Encompass_Preferred
Medchemexpress, HY-W008393 100mg (-)-Corey lactone diol CAS:32233-40-2 (-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4514 Supplier Medchemexpress LLC Supplier No. HYW008393100MG
Medchemexpress, HY-W008393 100mg (-)-Corey lactone diol CAS:32233-40-2 (-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.