Learn More
Medchemexpress LLC (-)-Corey lactone diol | 32233-40-2 | 172.18 | 1 ML
SDP
Catalog No. 5000447291
Encompass_Preferred
(-)-Corey lactone diol is the reduced form of Corey aldehyde, serving as a versatile drug intermediate for the synthesis of compounds such as Beraprost.
  • Solid, white to off-white appearance
  • Stable under recommended storage conditions
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage for solutions: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-004-47291 Supplier Medchemexpress LLC Supplier No. HYW00839310MM/1ML
(-)-Corey lactone diol is the reduced form of Corey aldehyde, serving as a versatile drug intermediate for the synthesis of compounds such as Beraprost.
  • Solid, white to off-white appearance
  • Stable under recommended storage conditions
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage for solutions: -80°C for 6 months; -20°C for 1 month