Learn More
Medchemexpress LLC (-)-Corey lactone diol | 32233-40-2 | 172.18 | 10 G
SDP
Catalog No. 5000449212
Encompass_Preferred
(-)-Corey lactone diol is the reduced form of Corey aldehyde, serving as a versatile drug intermediate. It is commonly used in the synthesis of compounds such as Beraprost.
  • Reduced form of Corey aldehyde
  • Used as a drug intermediate for synthesis
  • Appears as a solid
  • Purity 98.0%
  • White to off-white appearance
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-49212 Supplier Medchemexpress LLC Supplier No. HYW00839310G
(-)-Corey lactone diol is the reduced form of Corey aldehyde, serving as a versatile drug intermediate. It is commonly used in the synthesis of compounds such as Beraprost.
  • Reduced form of Corey aldehyde
  • Used as a drug intermediate for synthesis
  • Appears as a solid
  • Purity 98.0%
  • White to off-white appearance
  • Stable under recommended storage conditions
  • For research use only