Learn More
Medchemexpress LLC (-)-Corey lactone diol | 32233-40-2 | 172.18 | 500 MG
SDP
Catalog No. 5000445535
Encompass_Preferred
(-)-Corey lactone diol is an analytical standard and pharmaceutical intermediate. It is the reduced form of Corey aldehyde and serves as a building block in chemical synthesis. This product is intended for research and analytical applications, including use as a reference standard in HPLC, GC, and MS for qualitative, quantitative, and methodological research.
  • Analytical standard for research and analytical applications
  • Reduced form of corey aldehyde
  • Pharmaceutical intermediate in synthesis
  • Reference standard in HPLC, GC, and MS studies
  • Building block in chemical synthesis
  • Suitable as a laboratory chemical
Catalog No. 50-004-45535 Supplier Medchemexpress LLC Supplier No. HYW008393500MG
(-)-Corey lactone diol is an analytical standard and pharmaceutical intermediate. It is the reduced form of Corey aldehyde and serves as a building block in chemical synthesis. This product is intended for research and analytical applications, including use as a reference standard in HPLC, GC, and MS for qualitative, quantitative, and methodological research.
  • Analytical standard for research and analytical applications
  • Reduced form of corey aldehyde
  • Pharmaceutical intermediate in synthesis
  • Reference standard in HPLC, GC, and MS studies
  • Building block in chemical synthesis
  • Suitable as a laboratory chemical