Learn More
Cayman Chemical Corey Lactone Aldehyde Benzoate, Purity: ≥90%, CAS: 39746-01-5, 5 g
SDP
Catalog No. 502893592
Encompass_Preferred
Corey lactone aldehyde benzoate is a versatile, chiral intermediate used in the preparation of prostaglandins and prostaglandin analogs
Catalog No. 50-289-3592 Supplier Cayman Chemical Supplier No. 700305 G
Corey lactone aldehyde benzoate is a versatile, chiral intermediate used in the preparation of prostaglandins and prostaglandin analogs