Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Corilagin, a gallotannin | 23094-69-1 | 634.45 | 20 MG
SDP

Catalog No. 5000388590
Encompass_Preferred

Corilagin, a gallotannin, exhibits anti-tumor, anti-inflammatory, and hepatoprotective activities. It inhibits the activity of reverse transcriptase of RNA tumor viruses and the growth of *Staphylococcus aureus* (MIC of 25 μg/mL). Corilagin also demonstrates anti-tumor activity in hepatocellular carcinoma and ovarian cancer models, with low toxicity to normal cells and tissues.

  • Purity: 99.96%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants, Euphorbiaceae, Phyllanthus urinaria Linn.
  • Solubility in DMSO: 100 mg/mL (requires ultrasonic)
  • Exhibits anti-tumor activity
  • Exhibits anti-inflammatory activity
  • Exhibits hepatoprotective activities
  • Inhibits reverse transcriptase of RNA tumor viruses
  • Inhibits growth of Staphylococcus aureus
  • Shows low toxicity to normal cells and tissues

Catalog No. 50-003-88590 Supplier Medchemexpress LLC Supplier No. HYN046220MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corilagin, a gallotannin, exhibits anti-tumor, anti-inflammatory, and hepatoprotective activities. It inhibits the activity of reverse transcriptase of RNA tumor viruses and the growth of *Staphylococcus aureus* (MIC of 25 μg/mL). Corilagin also demonstrates anti-tumor activity in hepatocellular carcinoma and ovarian cancer models, with low toxicity to normal cells and tissues.

  • Purity: 99.96%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants, Euphorbiaceae, Phyllanthus urinaria Linn.
  • Solubility in DMSO: 100 mg/mL (requires ultrasonic)
  • Exhibits anti-tumor activity
  • Exhibits anti-inflammatory activity
  • Exhibits hepatoprotective activities
  • Inhibits reverse transcriptase of RNA tumor viruses
  • Inhibits growth of Staphylococcus aureus
  • Shows low toxicity to normal cells and tissues

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.