Learn More
Medchemexpress LLC HY-111048 10mg Medchemexpress, Corin CAS:1808113-09-8 Purity:>98%
SDP
Catalog No. 501968813
Encompass_Preferred
Medchemexpress, HY-111048 10mg Corin CAS:1808113-09-8 Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8813 Supplier Medchemexpress LLC Supplier No. HY11104810MG
Medchemexpress, HY-111048 10mg Corin CAS:1808113-09-8 Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.