Learn More
Medchemexpress LLC HY-111048 5mg Medchemexpress, Corin CAS:1808113-09-8 Purity:>98%
SDP
Catalog No. 501932353
Encompass_Preferred
Medchemexpress, HY-111048 5mg Corin CAS:1808113-09-8 Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC) , with a K i (inact) of 110 nM for LSD1 and an IC 50 of 147 nM for HDAC1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2353 Supplier Medchemexpress LLC Supplier No. HY1110485MG
Medchemexpress, HY-111048 5mg Corin CAS:1808113-09-8 Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC) , with a K i (inact) of 110 nM for LSD1 and an IC 50 of 147 nM for HDAC1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.