Learn More
Julabo USA Inc CORIO C-BT19 open heating circulator bath, 1kw heater, 19L plastic tank, to 100 C, 115V/60Hz.
SDP
Catalog No. NC1901283
Encompass_Preferred
JULABO CORIO™ C-BT19 Immersion Heating Circulator combination with transparent bath tank for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ C immersion circulator provides temperatures to 100 °C for internal bath applications for use with water only.Products include a quick connect clamp for positioning the CORIO™ C on tank rim. Polycarbonate bath tanks have volume graduations in L and Qt. 19L tank, 1kW heater, 115V/60hz.
Catalog No. NC1901283 Supplier Julabo USA Inc Supplier No. 9011319.22
JULABO CORIO™ C-BT19 Immersion Heating Circulator combination with transparent bath tank for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ C immersion circulator provides temperatures to 100 °C for internal bath applications for use with water only.Products include a quick connect clamp for positioning the CORIO™ C on tank rim. Polycarbonate bath tanks have volume graduations in L and Qt. 19L tank, 1kW heater, 115V/60hz.