Learn More
Julabo USA Inc CORIO C Immersion heating circulator, 1kW heating power, 100 C, 115V/60Hz
SDP
Catalog No. 501374381
Encompass_Preferred
JULABO CORIO™ C Immersion Circulators for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ C immersion circulator provides temperatures to 100 °C for internal bath applications up to 30 L with water only. The bath clamp accomodates mounting to rectangular and round bath tanks. 1kW heater, 115V/60Hz.
Catalog No. 50-137-4381 Supplier Julabo USA Inc Supplier No. 9011000.22
JULABO CORIO™ C Immersion Circulators for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ C immersion circulator provides temperatures to 100 °C for internal bath applications up to 30 L with water only. The bath clamp accomodates mounting to rectangular and round bath tanks. 1kW heater, 115V/60Hz.