Learn More
Julabo USA Inc CORIO CD-1200FW R1270 EA 115V
SDP
Catalog No. NC2747047
Encompass_Preferred
Water-cooled Refrigerated/heating circulator, JULABO, MAGIO CD-1200FW 115V/60Hz, R1270 Refrigerant; -50…+150 °C; 1kW heater. Cooling: 1.25kW at 20C; 1.1kW at 0C; 630W at -20C, 380W @ -30C, 210W at -40C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 93lbs
Catalog No. NC2747047 Supplier Julabo USA Inc Supplier No. 9012728.N1.02
Add to Cart
Add to Cart
Request a Quote
Water-cooled Refrigerated/heating circulator, JULABO, MAGIO CD-1200FW 115V/60Hz, R1270 Refrigerant; -50…+150 °C; 1kW heater. Cooling: 1.25kW at 20C; 1.1kW at 0C; 630W at -20C, 380W @ -30C, 210W at -40C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 93lbs