Learn More
Julabo USA Inc CORIO CD-200F R290 EA 115V
SDP
Catalog No. NC2747046
Encompass_Preferred
Refrigerated/heating circulator, JULABO, CORIO CD-200F, 230V/60Hz, R290 Refrigerant. -20…+150C. 2kW heater. Cooling power 220W at 20C 170W at 0C 130W at -10C 60W at -20C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 3...4L bath tank. 9.1 x 15.4 x 25.6 in. 57.3lbs
Catalog No. NC2747046 Supplier Julabo USA Inc Supplier No. 9012701.N1.13
Add to Cart
Add to Cart
Request a Quote
Refrigerated/heating circulator, JULABO, CORIO CD-200F, 230V/60Hz, R290 Refrigerant. -20…+150C. 2kW heater. Cooling power 220W at 20C 170W at 0C 130W at -10C 60W at -20C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 3...4L bath tank. 9.1 x 15.4 x 25.6 in. 57.3lbs