Learn More
Julabo USA Inc CORIO CD-800F R1270 EA 115V
SDP
Catalog No. NC2751004
Encompass_Preferred
Refrigerated/heating circulator, JULABO, CORIO CD-800F, 115V/60Hz, R1270 Refrigerant. -40…+150C. 1kW heater. Cooling power 850W at 20C 750W at 0C 400W at -20C 250W at -30C 110W at -40C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. 13 x 18.5 x 27.6 in. 93lbs
Catalog No. NC2751004 Supplier Julabo USA Inc Supplier No. 9012715.N1.02
Add to Cart
Add to Cart
Request a Quote
Refrigerated/heating circulator, JULABO, CORIO CD-800F, 115V/60Hz, R1270 Refrigerant. -40…+150C. 1kW heater. Cooling power 850W at 20C 750W at 0C 400W at -20C 250W at -30C 110W at -40C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. 13 x 18.5 x 27.6 in. 93lbs