Learn More
Julabo USA Inc CORIO CD Immersion circulator, 1kW heater, to 150 C; 115V/60Hz.
SDP
Catalog No. 501374383
Encompass_Preferred
JULABO CORIO™ Immersion Circulators for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ CD immersion circulator provides temperatures to 150 °C for internal bath applications up to 50 L with 1kW heating power. Single speed pump 15L/min to 5.1 PSI. Adjustable clamp. The CD includes a USB port for external PC control / communication and data-logging. With an optional pump set the CD can circulate to external applications with flow control via a manual lever. 115V/60Hz
Catalog No. 50-137-4383 Supplier Julabo USA Inc Supplier No. 9012000.02
JULABO CORIO™ Immersion Circulators for routine laboratory tasks. Units include bright LED display and easy operation. A quick connect bath clamp is included.The CORIO™ CD immersion circulator provides temperatures to 150 °C for internal bath applications up to 50 L with 1kW heating power. Single speed pump 15L/min to 5.1 PSI. Adjustable clamp. The CD includes a USB port for external PC control / communication and data-logging. With an optional pump set the CD can circulate to external applications with flow control via a manual lever. 115V/60Hz