Learn More
Julabo USA Inc CORIO CP-1200FW R1270 EA 115V
SDP
Catalog No. NC2747052
Encompass_Preferred
Water-cooled Refrigerated/heating circulator, JULABO, CORIO CP-1200FW, 115V/60Hz, R1270 Refrigerant; -50…+200C; 1kW heater. Cooling power: 1.24kW at 20C; 1.09kW at 0C; 620W at -20C; 370W at -30C; 200W at -40C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB, RS232 communication and USB data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 113.5lbs
Catalog No. NC2747052 Supplier Julabo USA Inc Supplier No. 9013728.N1.22
Add to Cart
Add to Cart
Request a Quote
Water-cooled Refrigerated/heating circulator, JULABO, CORIO CP-1200FW, 115V/60Hz, R1270 Refrigerant; -50…+200C; 1kW heater. Cooling power: 1.24kW at 20C; 1.09kW at 0C; 620W at -20C; 370W at -30C; 200W at -40C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB, RS232 communication and USB data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 113.5lbs