Learn More
Julabo USA Inc CORIO CP immersion heating circulator; +20…+200 °C operating range; ±0.02 °C stability; 1 kW heater, 8-27 L/min pump with 1.5-10.2 psi; adjustable clamp; applications to 50L; USB & RS232 communication
SDP
Catalog No. 501904647
Encompass_Preferred
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO™ CP immersion heating circulators are suitable for temperature applications up to 50L. Adjustable clamp makes installation easy. Operating range from +20…+200 °C.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.02 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.Class III (FL) according to DIN 12876-1.
Catalog No. 50-190-4647 Supplier Julabo USA Inc Supplier No. 9013000.22
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO™ CP immersion heating circulators are suitable for temperature applications up to 50L. Adjustable clamp makes installation easy. Operating range from +20…+200 °C.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.02 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.Class III (FL) according to DIN 12876-1.