Learn More
Medchemexpress LLC (±)-Coriolic acid | 18104-45-5 | 99.0% | 296.44 | 100 UG
SDP
Catalog No. 5000436536
Encompass_Preferred
(±)-Coriolic acid is one of the two racemic monohydroxy fatty acids resulting from the non-enzymatic oxidation of linoleic acid. It is the principal hydroxylated fatty acid in human psoriatic skin scales, with a mean concentration of 17 ng/mg.
  • Racemic monohydroxy fatty acid
  • Results from non-enzymatic oxidation of linoleic acid
  • Principal hydroxylated fatty acid in human psoriatic skin scales
  • Functions as an endogenous metabolite
  • Involved in metabolic enzyme/protease pathways
  • Stable under recommended storage conditions
  • Recommended for research use only
  • Store as a solution at -20°C for up to 2 years
Catalog No. 50-004-36536 Supplier Medchemexpress LLC Supplier No. HY113884A100UG
Add to Cart
Add to Cart
(±)-Coriolic acid is one of the two racemic monohydroxy fatty acids resulting from the non-enzymatic oxidation of linoleic acid. It is the principal hydroxylated fatty acid in human psoriatic skin scales, with a mean concentration of 17 ng/mg.
  • Racemic monohydroxy fatty acid
  • Results from non-enzymatic oxidation of linoleic acid
  • Principal hydroxylated fatty acid in human psoriatic skin scales
  • Functions as an endogenous metabolite
  • Involved in metabolic enzyme/protease pathways
  • Stable under recommended storage conditions
  • Recommended for research use only
  • Store as a solution at -20°C for up to 2 years