Learn More
Cayman Chemical Coriolis Compact air sampler
SDP
Catalog No. 502916837
Encompass_Preferred
The Coriolis Compact is a dry Cyclonic air sampler able to operate up to eight hours straight and compatible with multiple downstream analysis (NGS, qPCR, Culture).
Catalog No. 50-291-6837 Supplier Cayman Chemical Supplier No. 382841 EA
Add to Cart
Add to Cart
Request a Quote
The Coriolis Compact is a dry Cyclonic air sampler able to operate up to eight hours straight and compatible with multiple downstream analysis (NGS, qPCR, Culture).