Learn More
Electron Microscopy Sciences Cork Discs, 20 mm x 3 mm, 100/pk
SDP
Catalog No. 5031604
Encompass_Preferred
These discs are 20 mm diameter and are 3 mm thick. Perfect to be used with a cryostat.
Catalog No. 50-316-04 Supplier Electron Microscopy Sciences Supplier No. 63305
These discs are 20 mm diameter and are 3 mm thick. Perfect to be used with a cryostat.