Learn More
Andwin Scientific CORK SHEET 4 X12 3-1/8 THICK
SDP
Catalog No. NC9074469
Encompass_Preferred
The cork sheets are ideal for use to provide protection for workbench tops from heat, acids, and alkalis. This product can also be utilized as a dissecting pad. It measures 101 x 305 mm (4 x 12"). The sheet is 3 mm (1/8") thick. It is sold in a pack of 12.
Catalog No. NC9074469 Supplier Andwin Scientific Supplier No. 23420708
The cork sheets are ideal for use to provide protection for workbench tops from heat, acids, and alkalis. This product can also be utilized as a dissecting pad. It measures 101 x 305 mm (4 x 12"). The sheet is 3 mm (1/8") thick. It is sold in a pack of 12.