Learn More
ACCEGEN BIOTECH CORL311 Cell Line, ≥1.0 million/ frozen vial
Catalog No. NC3659658
Encompass
Human lung small cell carcinoma. COR-L311 has been isolated from a 63 year-old male patient with extensive disease and relapse after 3 cycles of chemotherapy treatment with eroposide, colchicine, amethopterin and vincristine.
Catalog No. NC3659658 Supplier ACCEGEN BIOTECH Supplier No. ABCTC0183
Add to Cart
Add to Cart
Human lung small cell carcinoma. COR-L311 has been isolated from a 63 year-old male patient with extensive disease and relapse after 3 cycles of chemotherapy treatment with eroposide, colchicine, amethopterin and vincristine.