Learn More
Apexbio Technology LLC CORM-3 475473-26-8 20mg
SDP
Catalog No. 502240214
Encompass_Preferred
CORM-3 (CAS 475473-26-8) is a water-soluble carbon monoxide-releasing molecule used in biomedical research It delivers controlled CO release activating soluble guanylate cyclase to mimic the anti-inflammatory effects observed with heme oxygenase activity In vitro studies have demonstrated that CORM-3 reduces PMN-induced CD54 expression on endothelial cells and exhibits vasoactive properties including modulation of vascular tone and blood pressure Additionally CORM-3 displays broad-spectrum antimicrobial activity by increasing membrane permeability and inhibiting terminal oxidases in pathogens This compound serves as a valuable tool for investigating CO s roles in cardiovascular inflammation vascular modulation and antimicrobial mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0214 Supplier Apexbio Technology LLC Supplier No. B306020
CORM-3 (CAS 475473-26-8) is a water-soluble carbon monoxide-releasing molecule used in biomedical research It delivers controlled CO release activating soluble guanylate cyclase to mimic the anti-inflammatory effects observed with heme oxygenase activity In vitro studies have demonstrated that CORM-3 reduces PMN-induced CD54 expression on endothelial cells and exhibits vasoactive properties including modulation of vascular tone and blood pressure Additionally CORM-3 displays broad-spectrum antimicrobial activity by increasing membrane permeability and inhibiting terminal oxidases in pathogens This compound serves as a valuable tool for investigating CO s roles in cardiovascular inflammation vascular modulation and antimicrobial mechanisms