Learn More
Medchemexpress LLC Corm-401 | 1001015-18-4 | 98.01% | 331.20 | 1 ML
SDP
Catalog No. 5000343264
Encompass_Preferred
CORM-401 is an oxidant-sensitive CO-releasing molecule that induces NO increase in the regulation of endothelial calcium signalling. It reduces TNF-α/CHX and H2O2-induced ROS production and uncouples mitochondrial respiration while inhibiting glycolysis.
  • Oxidant-sensitive CO-releasing molecule.
  • Induces NO increase in endothelial calcium signalling.
  • Reduces TNF-α/CHX and H2O2-induced ROS production and cell death.
  • Uncouples mitochondrial respiration and inhibits glycolysis.
Catalog No. 50-003-43264 Supplier Medchemexpress LLC Supplier No. HY10980450MG
CORM-401 is an oxidant-sensitive CO-releasing molecule that induces NO increase in the regulation of endothelial calcium signalling. It reduces TNF-α/CHX and H2O2-induced ROS production and uncouples mitochondrial respiration while inhibiting glycolysis.
  • Oxidant-sensitive CO-releasing molecule.
  • Induces NO increase in endothelial calcium signalling.
  • Reduces TNF-α/CHX and H2O2-induced ROS production and cell death.
  • Uncouples mitochondrial respiration and inhibits glycolysis.