Learn More
Medchemexpress LLC Corn oil | 8001-30-7 | 500 ML
SDP
Catalog No. 5000365851
Encompass_Preferred
Corn oil, extracted from the germ of corn, serves as a versatile carrier for agent molecules and a delivery vehicle for lipophilic substances in animal research. It is also commonly utilized as a feed additive. For optimal use, gavage administration is recommended, and proper storage is crucial to maintain product integrity and avoid contamination, especially for sensitive applications like IP injections.
  • Extracted from the germ of corn.
  • Functions as a carrier for agent molecules.
  • Acts as a delivery vehicle for lipophilic substances in animal research.
  • Commonly used as a feed additive.
  • Recommended for gavage administration.
  • Liquid form, colorless to light yellow.
  • Storage: -20°C for 3 years or 4°C for 2 years (pure form).
Catalog No. 50-003-65851 Supplier Medchemexpress LLC Supplier No. 500MLHYY1888
Corn oil, extracted from the germ of corn, serves as a versatile carrier for agent molecules and a delivery vehicle for lipophilic substances in animal research. It is also commonly utilized as a feed additive. For optimal use, gavage administration is recommended, and proper storage is crucial to maintain product integrity and avoid contamination, especially for sensitive applications like IP injections.
  • Extracted from the germ of corn.
  • Functions as a carrier for agent molecules.
  • Acts as a delivery vehicle for lipophilic substances in animal research.
  • Commonly used as a feed additive.
  • Recommended for gavage administration.
  • Liquid form, colorless to light yellow.
  • Storage: -20°C for 3 years or 4°C for 2 years (pure form).