Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Corn steep liquor | 66071-94-1 | 100 G
SDP

Catalog No. 5000283421
Encompass_Preferred

Corn steep liquor is a food-industry by-product supplied as a dried powder that serves as a nutrient-rich supplement for liquid culture media. It provides amino acids, vitamins, and minerals that support fungal mycelial growth and can increase production of secondary metabolites. Manufacturer storage and handling guidance should be followed for best results.

  • Used as a nutritional additive for fungal liquid culture media.
  • Enhances production of secondary metabolites and mycelial growth.
  • Provided as a yellow to brown powder form.
  • Available in multiple pack sizes for lab and scale-up use.
  • CAS number 66071-94-1 for substance identification.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Shipped at room temperature in continental US; conditions may vary elsewhere.

Catalog No. 50-002-83421 Supplier Medchemexpress LLC Supplier No. HYW133982100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corn steep liquor is a food-industry by-product supplied as a dried powder that serves as a nutrient-rich supplement for liquid culture media. It provides amino acids, vitamins, and minerals that support fungal mycelial growth and can increase production of secondary metabolites. Manufacturer storage and handling guidance should be followed for best results.

  • Used as a nutritional additive for fungal liquid culture media.
  • Enhances production of secondary metabolites and mycelial growth.
  • Provided as a yellow to brown powder form.
  • Available in multiple pack sizes for lab and scale-up use.
  • CAS number 66071-94-1 for substance identification.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Shipped at room temperature in continental US; conditions may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.