Learn More
Haematologic Technologies Inc CORN TRYPSIN INHIBITOR 1MG
Supplier:  Haematologic Technologies Inc CTI01
Encompass
Inhibitor, Corn Trypsin Inhibitor, Purified from corn, 1 milligram, Use: Inhibitor of trypsin and Factor XIIa, Research use only, Supplied in Tris buffered saline, store at -80 Celsius.
Catalog No. 50-883-459