Learn More
Sciencell Research Laboratories CORNEAL EPITHELIAL CELL MEDIUM
Catalog No. NC2069905
Encompass
CEpiCM consists of 500 ml of basal medium, 5 ml of corneal epithelial cell growth supplement (CEpiCGS, Cat. No. 6552) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).
Catalog No. NC2069905 Supplier Sciencell Research Laboratories Supplier No. 6511
CEpiCM consists of 500 ml of basal medium, 5 ml of corneal epithelial cell growth supplement (CEpiCGS, Cat. No. 6552) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).