Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 1536-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GSA_VA

Catalog No. 13701101 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-701-101 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-701-101 Supplier Corning™ Supplier No. 3839BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.

Order Info

Packaged in bags of 10

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Lid With Lid
Array 32 x 48
Packaging 10/Pk., 50/Cs.
Binding Type Poly-D-Lysine
Color Black, Clear Bottom
Product Type Cell Culture Plate
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
For Use With (Application) For fluorescent and luminescent assays
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.