Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellBIND™ 1536-Well, CellBIND-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 12456702 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-456-702 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-456-702 Supplier Corning™ Supplier No. 4563
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Lid Without Lid
Culture Area 0.03 cm2
Array 32 x 48
Binding Type Corning CellBIND Surface
Color Black, Clear Bottom
For Use With (Equipment) Automated dispensers
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Sterile
Surface Treatment CellBIND
Well Shape Flat Bottom (F-well)
For Use With (Application) For both top and bottom reading instruments
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 20/Pk.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.