Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 1536-Well, CellBIND-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07200676 Shop All Corning Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-676 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-676 Supplier Corning™ Supplier No. 3731BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Color:White; Sterility:Sterile; Lid:With Lid

  • Conform to standard microplate footprint and dimensions
  • Flood reservoir on four sides to reduce instrument contamination
  • CellBIND treated
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Binding Property CellCIND treated
Format Standard
Lid With Lid
Array 32 x 48
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 1536
Sterility Sterile
Surface Treatment CellBIND
Well Shape Flat Bottom (F-well)
For Use With (Application) High throughput screening
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.