Learn More
Corning™ 1536-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 12456705 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
12-456-705 Black
1 options
Catalog No. 12-456-705 Supplier Corning™ Supplier No. 4566
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Color:Black; Sterility:Non-sterile; Lid:Without Lid
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Lid Without Lid
Culture Area 0.03 cm2
Array 32 x 48
Binding Type TC-Treated
Color Black
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 12.5 μL
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 10/Pk.
Well Color Black
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov