Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07201321 Shop All Corning Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07201321 White
1 options
Catalog No. 07201321 Supplier Corning™ Supplier No. 3572
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Array 16 x 24
Color White
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.