Learn More
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
Specifications
| Lid | Without Lid |
| Array | 16 x 24 |
| Binding Type | High Binding |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
