Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07200345 Shop All Corning Products
Change view
Click to view available options
Quantity:
50/Cs.
Catalog No. Quantity
07-200-345 50/Cs.
1 options
Catalog No. 07-200-345 Supplier Corning™ Supplier No. 3570BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background fluorescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112μL
  • Working volume: 20μL to 80μL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Cs.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov