Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE

Catalog No. 07200891 Shop All Corning Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-891 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-891 Supplier Corning™ Supplier No. 3820
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.

Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

TRUSTED_SUSTAINABILITY

Specifications

Lid Without Lid
Description (NBS) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions.
Array 16 x 24
Binding Type Non Binding
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Non-sterile
Surface Treatment Non-binding
Total Volume (Metric) Well 50 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.