Learn More
Description
- Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
- Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
- Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
- Use at temperature range of 4° to 37°C (39° to 99°F)
- Flat-bottom wells
- Compatible with a wide range of plate readers, liquid handlers, and robotic systems
Generic Bar Coded Plates
- Code 128 bar code format ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified
- Labels applied to all four sides of the plate No sequence number duplicates
Specifications
Specifications
| Barcode | Includes Barcode |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Black |
| For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Non-sterile |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
