Learn More
Corning™ 96-Well, High Binding, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07200400 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-200-400 Clear
1 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-400 Supplier Corning™ Supplier No. 72592
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Sold 100 per case.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
No. of Wells 96
Sterility Non-sterile
Surface Treatment High Binding
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov