Learn More
Corning™ 96-Well, Multi-Treated, Flat-Bottom Microplate
Catalog No. 07201190 Shop All Corning Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
07-201-190 Sterile
1 options
Catalog No. 07-201-190 Supplier Corning™ Supplier No. 3823
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340nm
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • Nonreversable lids with condensation rings to reduce contamination
  • Six different surface treatments on a single plate ((Collagen I, Fibronectin, Gelatin, Laminin, Poly-D-Lysine, TC-treated))
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Packaging 10/Pk.
Color Black, Clear Bottom
Product Type multiPlate
Material Polystyrene
Volume (Metric) Well 100 to 200 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Multi-treated
Well Shape Round, Flat Bottom (F-well)
No. per Case 10/Cs.
No. per Pack 1/Pk.
Quantity 10/Cs.
Well Color Clear
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov