Learn More
Corning™ 96-Well Half Area High Content Imaging Glass Bottom Microplate
GREENER_CHOICE
Catalog No. 13701102 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
13-701-102 Black
1 options
This item is not returnable. View return policy
Catalog No. 13-701-102 Supplier Corning™ Supplier No. 4580
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
High optical-quality glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems
High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
  • Half area 96-well microplate reduces reagent consumption.
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 8 x 12
Color Black
Material Cycloolefin Copolymer
No. of Wells 96
Sterility Sterile
For Use With (Application) Provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth
No. per Case 10/Cs.
No. per Pack 1/Pk.
WARNING: Cancer - www.P65Warnings.ca.gov