Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well Microplate

Catalog No. 07201207 Shop All Corning Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-201-207 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-201-207 Supplier Corning™ Supplier No. 3995

missing translation for 'validMainframeMsg'

Designed to reduce well-to-well crosstalk

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Lid Without Lid
    Array 8 x 12
    Packaging Bags of 5
    Color White
    Product Type 96-well Assay Microplate
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 25/Cs.
    No. per Pack 5/Pk.
    Quantity 25/Cs.
    Show More Show Less
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.