Learn More
Waters Corp Corning Well Plate, 96 Well, Half Vol, Black Polystyrene
Catalog No. NC2254673 Shop All Waters Corporation Products
Encompass_Preferred
A 96 well plate designed for use with a 9 mm insert, featuring a minimum sample volume of 100 μL and a temperature range of 0°C to 60°C. The well wall is made of black polystyrene, and the well bottom is clear polystyrene.
  • Designed for use with a 9 mm insert.
  • Minimum sample volume of 100 μL.
  • Temperature range of 0°C to 60°C.
  • Well wall material is black polystyrene.
  • Well bottom material is clear polystyrene.
  • Compatible with Plate Reader 3 and 4.
  • Can be used with SLS when using oil or sealing tape for evaporation control.
  • Compatible with the DynaPro Plate Reader III.
Catalog No. NC2254673 Supplier Waters Corp Supplier No. P880209602
A 96 well plate designed for use with a 9 mm insert, featuring a minimum sample volume of 100 μL and a temperature range of 0°C to 60°C. The well wall is made of black polystyrene, and the well bottom is clear polystyrene.
  • Designed for use with a 9 mm insert.
  • Minimum sample volume of 100 μL.
  • Temperature range of 0°C to 60°C.
  • Well wall material is black polystyrene.
  • Well bottom material is clear polystyrene.
  • Compatible with Plate Reader 3 and 4.
  • Can be used with SLS when using oil or sealing tape for evaporation control.
  • Compatible with the DynaPro Plate Reader III.