Learn More
Corning™ 96-Well Round Bottom Polystyrene Ulta-Low Attachment Microplate
GSA_VAGREENER_CHOICE
Catalog No. 15100175 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
15-100-175 Black
1 options
Catalog No. 15-100-175 Supplier Corning™ Supplier No. 4591
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Ulta-Low attachment surface with covalently bonded hyrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Array 8 x 12
    Color Black
    Material Polystyrene
    Volume (Metric) Well 360 μL
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Well Shape Round
    No. per Case 24/Cs.
    No. per Pack 1/Pk.