Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07200729 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-729 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-729 Supplier Corning™ Supplier No. 3614
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Lid Without Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Color Black, Clear Bottom
    Product Type Microplate
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Spectroscopy, polarization and imaging
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.