Learn More
Corning™ Adapters For Round-Bottom Centrifuge Tubes
Catalog No. 0556656 Shop All Corning Products
Change view
Click to view available options
Adapters, For Tube Cat. No.: 05-566-55
Catalog No.
05-566-56 Adapters, For Tube Cat. No.: 05-566-55
1 options
Catalog No. 05-566-56 Supplier Corning™ Supplier No. 8445AO
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Requires
For use in 05-375-80, also requires Thermo Scientific 6650E adapters.
  • Adapters For Corex™ II Tubes. Required when centrifuging selected Corex II round-bottom centrifuge tubes. Made of rubber.