Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Empty Racks With Lids for 96 Screw Capped or Thermoplastic Elastomers (TPE) Capped Storage Tubes

Catalog No. 07200518 Shop All Corning Products
Change view
Click to view available options
Positions:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
07-200-518 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-518 Supplier Corning™ Supplier No. 8508
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

For use with Mfr. Nos. 6500 and 8502

Designed for use at temperatures ranging from -100°C to +110°C.

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Description Empty Racks
Holds 96 Screw Capped or Thermoplastic Elastomers (TPE) Capped Storage Tubes
Material Polypropylene
Positions 96
Sterility Non-sterile
For Use With (Equipment) Mfr. Nos. 6500 and 8502
No. per Case 2 Pk./Cs.
No. per Pack 5/Pk.
Temperature (Metric) Range −100° to 110°C
Dimensions (L x W) 127.8 x 85.5 mm
Includes Locking Lid
Array 8 x 12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.