Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 08774297 Shop All Corning Products
Change view
Click to view available options
Packaging:
5/Cs
5/Pk., 50/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
08-774-297 5/Pk., 50/Cs.
08-774-294 5/Cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-774-297 Supplier Corning™ Supplier No. 356519
Only null left
Add to Cart
Add to Cart

Color: White; Sterility: Sterile; Lid: With Lid

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Culture Area 0.25 cm2
    Array 8 x 12
    Packaging 5/Pk., 50/Cs.
    Color White
    Product Type 96-well Collagen I Multiwell Microplate
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Collagen Type I
    Well Shape Round, Flat Bottom (F-well)
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Quantity 50/Cs.
    Source Rat Tail
    Well Color Opaque
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.