Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate

Catalog No. 15100160 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-100-160 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-100-160 Supplier Corning™ Supplier No. 4582

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid

High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
  • Half area 96-well microplate reduces reagent consumption.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Lid With Lid
Array 8 x 12
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Collagen Type I
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
No. per Case 10/Cs.
No. per Pack 1/Pk.
Quantity 10/Cs.
Source Rat Tail
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.