Learn More
Corning™ BioCoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate
Shop All Corning ProductsDescription
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 μm is well suited for imaging microscopy.
- Bottom flatness <50 μm to ensure planarity for imaging devices
- Low background fluorescence and minimal cross-talk provide the highest possible optical quality for cell-based assays.
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | ANSI, SBS |
| Lid | With Lid |
| Array | 16 x 24 |
| Color | Black |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Surface Treatment | Collagen Type I |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.